Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHACTR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199505.1 | NP_001186434.1 | 7 | 1145 | ||
NM_001199506.1 | NP_001186435.1 | 7 | 1140 | |||
NM_001281507.1 | NP_001268436.1 | 7 | 1244 | |||
NM_080672.4 | NP_542403.1 | 7 | 1622 | |||
NM_183244.1 | NP_899067.1 | 7 | 1265 | |||
NM_183246.1 | NP_899069.1 | 6 | 1055 | |||
XM_011528525.2 | XP_011526827.1 | 7 | 1251 | |||
XM_011528526.2 | XP_011526828.1 | 6 | 1057 | |||
XM_017027626.1 | XP_016883115.1 | 7 | 1354 | |||
XM_017027627.1 | XP_016883116.1 | 7 | 1299 | |||
XM_017027628.1 | XP_016883117.1 | 6 | 947 | |||
XM_017027629.1 | XP_016883118.1 | 6 | 1090 | |||
XM_017027630.1 | XP_016883119.1 | 6 | 918 | |||
XM_017027631.1 | XP_016883120.1 | 7 | 1181 | |||
GenBank mRNA | AB098521.1 | - | 7 | 1265 | ||
AB098522.1 | - | 6 | 1055 | |||
AJ311122.2 | - | 7 | 1621 | |||
AJ617581.1 | - | 7 | 1140 | |||
AK091632.1 | - | 7 | 1267 | |||
AK098788.1 | - | 7 | 1145 | |||
AK295459.1 | - | 7 | 1244 | |||
AK314493.1 | - | 7 | 1352 | |||
AK316047.1 | - | 7 | 1137 | |||
BC108303.1 | - | 7 | 1622 | 4179728 | ||
BC117362.1 | - | 7 | 1510 | 40125913 | ||
BC117364.1 | - | 7 | 1513 | 40125915 |