Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037288.1 | NP_001032365.1 | 3 | 774 | ||
NM_001288744.1 | NP_001275673.1 | 3 | 714 | |||
NM_001288745.1 | NP_001275674.1 | 2 | 621 | |||
NM_001288746.1 | NP_001275675.1 | 5 | 1068 | |||
NM_138460.2 | NP_612469.1 | 4 | 867 | |||
XM_017020953.1 | XP_016876442.1 | 5 | 853 | |||
XM_017020954.1 | XP_016876443.1 | 5 | 730 | |||
GenBank mRNA | AF479262.2 | - | 4 | 868 | ||
AF527413.1 | - | 5 | 851 | |||
AF527414.1 | - | 4 | 498 | |||
AF527948.2 | - | 2 | 392 | |||
AF527949.1 | - | 3 | 485 | |||
AK094840.1 | - | 3 | 1599 | |||
AY820135.1 | - | 3 | 330 | |||
BC013109.1 | - | 4 | 612 | 4214683 |