Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERCC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000082.3 | NP_000073.1 | 2, 3 | 237 | ||
NM_001007234.2 | NP_001007235.1 | 2, 3 | 237 | |||
NM_001290285.1 | NP_001277214.1 | 2, 3 | 237 | |||
GenBank mRNA | AK290726.1 | - | 2, 3 | 217 | ||
AK294856.1 | - | 2, 3 | 218 | |||
AK304610.1 | - | 2, 3 | 167 | |||
AK314511.1 | - | 2, 3 | 210 | |||
AL691658.1 | - | 2, 3 | 224 | |||
BC009793.1 | - | 2, 3 | 214 | 3683434 | ||
BT020021.1 | - | 2, 3 | 167 | |||
CR536563.1 | - | 2, 3 | 167 | |||
DA169067.1 | - | 2, 3 | 237 | |||
KU177979.1 | - | 2, 3 | 167 | |||
U28413.1 | - | 2, 3 | 203 |