Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040118.2 | NP_001035207.1 | 18 | 2708 | ||
NM_001135190.1 | NP_001128662.1 | 15 | 2336 | |||
NM_015242.4 | NP_056057.2 | 16 | 2519 | |||
GenBank mRNA | AB018325.2 | - | 16 | 2496 | ||
AF411983.1 | - | 12 | 1577 | |||
AJ621557.1 | - | 16 | 2519 | |||
AK160383.1 | - | 15 | 2858 | |||
AY049732.1 | - | 13 | 2356 | |||
AY553630.1 | - | 18 | 2723 | |||
BC056401.1 | - | 4 | 362 | |||
BC140792.1 | - | 15 | 2006 | 9021660 | ||
BC144562.1 | - | 16 | 2189 | 9053090 |