Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024647.3 | NP_001019818.1 | 4, 5 | 612 | ||
NM_001278402.1 | NP_001265331.1 | 4, 5 | 527 | |||
NM_022456.4 | NP_071901.2 | 6, 7 | 1263 | |||
NM_175623.3 | NP_783322.1 | 6, 7 | 1397 | |||
NM_175624.3 | NP_783323.1 | 6, 7 | 1263 | |||
NM_175625.3 | NP_783324.1 | 6, 7 | 1397 | |||
XM_006719226.3 | XP_006719289.3 | 6, 7 | 1386 | |||
XM_006719227.3 | XP_006719290.3 | 6, 7 | 1059 | |||
XM_017018773.1 | XP_016874262.1 | 6, 7 | 1021 | |||
XM_017018774.1 | XP_016874263.1 | 8, 9 | 1274 | |||
XM_017018776.1 | XP_016874265.1 | 7, 8 | 1146 | |||
XM_017018777.1 | XP_016874266.1 | 7, 8 | 1565 | |||
GenBank mRNA | AJ312896.1 | - | 6, 7 | 1087 | ||
AJ312897.1 | - | 6, 7 | 1379 | |||
AJ312898.1 | - | 6, 7 | 1087 | |||
AJ312899.1 | - | 6, 7 | 1379 | |||
AK026201.1 | - | 6, 7 | 1087 | |||
AK027566.1 | - | 4, 5 | 523 | |||
AK027671.1 | - | 4, 5 | 513 | |||
AK057844.1 | - | 6, 7 | 992 | |||
AK225441.1 | - | 6, 7 | 1087 | |||
AK314547.1 | - | 6, 7 | 1098 | |||
BC015548.1 | - | 4, 5 | 590 | 3846338 | ||
DN912041.1 | - | 4, 5 | 612 |