Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054112.2 | NP_473453.1 | 2 | 317 | ||
GenBank mRNA | AF347073.1 | - | 2 | 310 | ||
AF529415.1 | - | 2 | 284 | |||
AY122471.1 | - | 1 | 219 | |||
BC117378.1 | - | 2 | 308 | 40125929 |