Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002924.2 | NP_001002924.2 | 7 | 956 | ||
NM_001284.3 | NP_001275.1 | 6 | 920 | |||
NM_001318090.1 | NP_001305019.1 | 4 | 716 | |||
NM_001318091.1 | NP_001305020.1 | 5 | 848 | |||
NM_001318093.1 | NP_001305022.1 | 7 | 961 | |||
NM_001318094.1 | NP_001305023.1 | 5 | 812 | |||
GenBank mRNA | AK124911.1 | - | 1 | 1621 | ||
AK126876.1 | - | 1 | 4271 | |||
AK311843.1 | - | 6 | 488 | |||
BC000804.1 | - | 6 | 572 | 3452302 | ||
BC012614.1 | - | 7 | 569 | |||
BI600357.1 | - | 6 | 608 | |||
BT009833.1 | - | 6 | 488 | |||
BX394598.2 | - | 5 | 391 | |||
CN411347.1 | - | 5 | 529 | |||
CR450341.1 | - | 6 | 488 | |||
D63643.1 | - | 6 | 530 | |||
DB483797.1 | - | 4 | 402 | |||
U91932.1 | - | 6 | 573 | |||
X99458.1 | - | 6 | 518 |