Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145806.3 | NP_665805.2 | 5, 6 | 725 | ||
NR_130127.1 | 5, 6 | 666 | ||||
GenBank mRNA | BC015139.1 | - | 5, 6 | 715 | ||
BC019897.1 | - | 5, 6 | 695 | 5123355 |