Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318189.1 | NP_001305118.1 | 14 | 1498 | ||
NM_144587.3 | NP_653188.2 | 14 | 1495 | |||
XM_011539239.2 | XP_011537541.1 | 13 | 1929 | |||
XM_011539240.2 | XP_011537542.1 | 13 | 1904 | |||
XM_011539241.2 | XP_011537543.1 | 12 | 1847 | |||
XM_011539242.2 | XP_011537544.1 | 11 | 1728 | |||
XM_017015636.1 | XP_016871125.1 | 13 | 2578 | |||
XM_017015637.1 | XP_016871126.1 | 10 | 2203 | |||
GenBank mRNA | AK058088.1 | - | 14 | 1485 | ||
BC109261.1 | - | 14 | 1469 | 40025404 | ||
BC109262.1 | - | 14 | 1469 | 40025414 | ||
DB338661.1 | - | 3 | 379 |