Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LARGE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300721.1 | NP_001287650.1 | 5 | 649 | ||
NM_001300722.1 | NP_001287651.1 | 6 | 576 | |||
NM_152312.4 | NP_689525.3 | 5 | 669 | |||
XM_005252787.2 | XP_005252844.1 | 1 | 42 | |||
XM_006718140.1 | XP_006718203.1 | 6 | 556 | |||
XM_006718141.3 | XP_006718204.1 | 5 | 630 | |||
XM_011519886.1 | XP_011518188.1 | 5 | 630 | |||
XM_011519888.1 | XP_011518190.1 | 5 | 630 | |||
XM_011519889.1 | XP_011518191.1 | 6 | 556 | |||
XM_011519890.1 | XP_011518192.1 | 5 | 630 | |||
XM_011519891.1 | XP_011518193.1 | 5 | 630 | |||
XM_011519892.2 | XP_011518194.1 | 5 | 630 | |||
XM_011519893.1 | XP_011518195.1 | 5 | 630 | |||
XM_017017173.1 | XP_016872662.1 | 4 | 723 | |||
GenBank mRNA | AK055829.1 | - | 6 | 576 | ||
AK092526.1 | - | 2 | 170 | |||
AK096021.1 | - | 5 | 645 | |||
BC037291.2 | - | 5 | 633 | 4821103 |