Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHEBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303126.1 | NP_001290055.1 | 3 | 447 | ||
NM_144593.2 | NP_653194.1 | 3 | 379 | |||
XM_017018794.1 | XP_016874283.1 | 3 | 1036 | |||
XM_017018795.1 | XP_016874284.1 | 3 | 1104 | |||
GenBank mRNA | AK098663.1 | - | 3 | 378 | ||
AY327412.1 | - | 3 | 379 | |||
AY509932.1 | - | 3 | 445 | |||
BC027482.1 | - | 3 | 346 | 4396827 |