Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001834.3 | NP_001825.1 | 1 | 309 | ||
NM_007097.3 | NP_009028.1 | 1 | 309 | |||
NR_045724.1 | 1 | 309 | ||||
XM_017009027.1 | XP_016864516.1 | 1 | 355 | |||
GenBank mRNA | AK300200.1 | - | 1 | 279 | ||
BC006332.2 | - | 1 | 247 | 4299637 | ||
BC006457.1 | - | 1 | 233 | 3538261 | ||
BM761740.1 | - | 1 | 279 | |||
BT007028.1 | - | 1 | 104 | |||
CN409237.1 | - | 1 | 311 | |||
CR536577.1 | - | 1 | 104 | |||
M20469.1 | - | 1 | 276 | |||
M20470.1 | - | 1 | 243 |