Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017523.1 | NP_001017523.1 | 15 | 1850 | ||
NM_001018072.1 | NP_001018082.1 | 17 | 3710 | |||
XM_005268645.3 | XP_005268702.1 | 17 | 2078 | |||
XM_011537909.2 | XP_011536211.1 | 17 | 2633 | |||
XM_011537910.2 | XP_011536212.1 | 17 | 2587 | |||
XM_017018807.1 | XP_016874296.1 | 16 | 4860 | |||
GenBank mRNA | AK091276.1 | - | 7 | 685 | ||
AK122818.1 | - | 17 | 2147 | |||
AK127012.1 | - | 14 | 1620 | |||
AK127295.1 | - | 15 | 1850 | |||
AK128445.1 | - | 9 | 1089 | |||
AY373588.1 | - | 13 | 1544 | |||
BC093627.1 | - | 6 | 516 | |||
BC093629.1 | - | 6 | 516 | |||
BC101561.1 | - | 6 | 516 | |||
BC101563.1 | - | 6 | 516 |