Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172193.2 | NP_751943.1 | 11, 12 | 1080 | ||
XM_011536422.2 | XP_011534724.1 | 11, 12 | 1003 | |||
GenBank mRNA | AF111806.1 | - | 11, 12 | 1018 | ||
AK127202.1 | - | 11, 12 | 1003 | |||
AL833437.1 | - | 6, 7 | 510 | |||
BC031270.1 | - | 11, 12 | 1043 | |||
BC101595.1 | - | 11, 12 | 1080 | 8069101 | ||
BC101597.1 | - | 11, 12 | 1080 | 8069103 |