Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC51B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178859.3 | NP_849190.2 | 4 | 688 | ||
XM_005254159.4 | XP_005254216.1 | 4 | 663 | |||
GenBank mRNA | AK129601.1 | - | 4 | 591 | ||
AY194242.1 | - | 3 | 397 | |||
BC103842.1 | - | 3 | 394 | 40002450 | ||
BC103843.1 | - | 3 | 394 | 40002452 | ||
BC103844.1 | - | 3 | 394 | 40002453 |