Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164680.1 | NP_001158152.1 | 3 | 429 | ||
NM_001837.3 | NP_001828.1 | 3 | 448 | |||
NM_178328.1 | NP_847898.1 | 3 | 438 | |||
NM_178329.2 | NP_847899.1 | 2 | 369 | |||
XM_006712960.3 | XP_006713023.1 | 3 | 475 | |||
XM_011533335.2 | XP_011531637.1 | 3 | 555 | |||
XM_017005685.1 | XP_016861174.1 | 3 | 1358 | |||
XM_017005686.1 | XP_016861175.1 | 4 | 1539 | |||
GenBank mRNA | AF026535.1 | - | 1 | 200 | ||
AF262299.1 | - | 2 | 319 | |||
AF262301.1 | - | 3 | 359 | |||
AF262302.1 | - | 3 | 369 | |||
BC110297.1 | - | 2 | 278 | 5212541 | ||
BC130318.1 | - | 2 | 280 | 40146349 | ||
BC130320.1 | - | 2 | 280 | 40146351 | ||
U28694.1 | - | 1 | 231 |