Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMKLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142343.1 | NP_001135815.1 | 4 | 629 | ||
NM_001142344.1 | NP_001135816.1 | 3 | 416 | |||
NM_001142345.1 | NP_001135817.1 | 3 | 243 | |||
NM_004072.2 | NP_004063.1 | 3 | 553 | |||
XM_017018820.1 | XP_016874309.1 | 2 | 371 | |||
GenBank mRNA | AB112777.1 | - | 1 | 46 | ||
AK291800.1 | - | 3 | 410 | |||
AY497547.1 | - | 1 | 46 | |||
BC106927.2 | - | 4 | 443 | 40033186 | ||
BC106928.1 | - | 4 | 443 | 40033187 | ||
BP328503.1 | - | 3 | 243 | |||
BT019556.1 | - | 1 | 46 | |||
BT019557.1 | - | 1 | 46 | |||
DA466082.1 | - | 3 | 416 | |||
DA939526.1 | - | 3 | 224 | |||
U79526.1 | - | 3 | 406 | |||
U79527.1 | - | 3 | 559 |
Set Membership: |
G-protein coupled receptor |