Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTB4R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143919.2 | NP_001137391.1 | 2 | 747 | ||
NM_181657.3 | NP_858043.1 | 2 | 2355 | |||
GenBank mRNA | BC004545.2 | - | 2 | 758 | 3946189 | |
BT007267.1 | - | 1 | 426 | |||
D89078.1 | - | 2 | 2143 | |||
D89079.1 | - | 2 | 747 | |||
U41070.1 | - | 2 | 730 | |||
X98356.1 | - | 2 | 814 |
Set Membership: |
G-protein coupled receptor |