Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152344.3 | NP_689557.1 | 3, 4 | 579 | ||
GenBank mRNA | AK055218.1 | - | 2, 3 | 307 | ||
AL832076.1 | - | 1 | 268 | |||
BC016791.2 | - | 1 | 80 | |||
BC044587.1 | - | 2, 3 | 307 | 4794614 | ||
BC105288.1 | - | 2, 3 | 321 | 6385345 |