Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB5D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144611.3 | NP_653212.1 | 1 | 812 | ||
XM_017024183.1 | XP_016879672.1 | 1 | 506 | |||
GenBank mRNA | AK172844.1 | - | 1 | 278 | ||
AK313088.1 | - | 1 | 283 | |||
BC020263.1 | - | 1 | 264 | 4877960 | ||
BC051697.1 | - | 1 | 724 | 5190840 |