Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139162.3 | NP_631901.2 | 3, 4 | 527 | ||
NM_148886.1 | NP_683684.2 | 3, 4 | 375 | |||
XM_017024190.1 | XP_016879679.1 | 3, 4 | 872 | |||
GenBank mRNA | AF467443.1 | - | 3, 4 | 375 | ||
AK056165.1 | - | 3, 4 | 506 | |||
BC014973.1 | - | 3, 4 | 375 | 4859126 | ||
DB183618.1 | - | 3, 4 | 533 |