Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042450.2 | NP_001035915.1 | 2 | 176 | ||
NM_001270648.1 | NP_001257577.1 | 2 | 176 | |||
NM_001270649.1 | NP_001257578.1 | 2 | 176 | |||
NM_001282417.1 | NP_001269346.1 | 2 | 207 | |||
NM_152351.4 | NP_689564.3 | 2 | 176 | |||
XM_011523652.1 | XP_011521954.1 | 2 | 332 | |||
XM_011523653.2 | XP_011521955.1 | 2 | 337 | |||
XM_017024191.1 | XP_016879680.1 | 2 | 333 | |||
GenBank mRNA | AK057946.1 | - | 2 | 176 | ||
AK298345.1 | - | 2 | 162 | |||
BC034380.1 | - | 2 | 121 | |||
BC039868.1 | - | 2 | 224 | 4157703 | ||
BC062617.1 | - | 2 | 130 | |||
BP332461.1 | - | 2 | 176 | |||
DR003789.1 | - | 2 | 176 |