Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOXHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144612.6 | NP_653213.6 | 15 | 2031 | ||
XM_011525804.2 | XP_011524106.1 | 3 | 265 | |||
XM_017025540.1 | XP_016881029.1 | 7 | 1061 | |||
XM_017025541.1 | XP_016881030.1 | 8 | 1148 | |||
XM_017025542.1 | XP_016881031.1 | 8 | 1148 | |||
XM_017025543.1 | XP_016881032.1 | 8 | 1148 | |||
XM_017025544.1 | XP_016881033.1 | 8 | 1148 | |||
XM_017025545.1 | XP_016881034.1 | 8 | 1148 | |||
XM_017025546.1 | XP_016881035.1 | 8 | 1148 | |||
XM_017025547.1 | XP_016881036.1 | 8 | 1148 | |||
XM_017025548.1 | XP_016881037.1 | 15 | 2113 | |||
GenBank mRNA | AK127869.1 | - | 8 | 1344 |