Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOXHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145472.2 | NP_001138944.1 | 12 | 1638 | ||
NM_001308013.1 | NP_001294942.1 | 10 | 1292 | |||
NM_144612.6 | NP_653213.6 | 30 | 4562 | |||
XM_006722388.3 | XP_006722451.1 | 11 | 1472 | |||
XM_006722389.3 | XP_006722452.1 | 12 | 1648 | |||
XM_006722390.3 | XP_006722453.1 | 12 | 1716 | |||
XM_006722391.3 | XP_006722454.1 | 11 | 1472 | |||
XM_011525804.2 | XP_011524106.1 | 18 | 2796 | |||
XM_011525807.2 | XP_011524109.1 | 9 | 1182 | |||
XM_017025540.1 | XP_016881029.1 | 22 | 3592 | |||
XM_017025541.1 | XP_016881030.1 | 23 | 3679 | |||
XM_017025542.1 | XP_016881031.1 | 23 | 3679 | |||
XM_017025543.1 | XP_016881032.1 | 23 | 3679 | |||
XM_017025544.1 | XP_016881033.1 | 23 | 3679 | |||
XM_017025545.1 | XP_016881034.1 | 23 | 3679 | |||
XM_017025546.1 | XP_016881035.1 | 24 | 3875 | |||
XM_017025547.1 | XP_016881036.1 | 22 | 3524 | |||
XM_017025548.1 | XP_016881037.1 | 28 | 4026 | |||
GenBank mRNA | AK127869.1 | - | 23 | 3875 | ||
AK302484.1 | - | 11 | 1469 | |||
BC041860.1 | - | 12 | 1620 | 5271046 | ||
BX647229.1 | - | 10 | 1292 |