Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102597.2 | NP_001096067.2 | 1, 2 | 62 | ||
NM_001102598.2 | NP_001096068.2 | 1, 2 | 62 | |||
NM_001102599.2 | NP_001096069.2 | 1, 2 | 62 | |||
NM_001102600.2 | NP_001096070.2 | 1, 2 | 62 | |||
GenBank mRNA | AY358129.1 | - | 1, 2 | 325 |