Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF440 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152357.2 | NP_689570.2 | 4 | 1833 | ||
XM_005259731.3 | XP_005259788.1 | 4 | 1893 | |||
XM_017026253.1 | XP_016881742.1 | 3 | 2218 | |||
XM_017026254.1 | XP_016881743.1 | 3 | 1711 | |||
GenBank mRNA | AK095252.1 | - | 4 | 1833 | ||
BC035760.1 | - | 4 | 1816 | 5556302 |