Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IZUMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152358.2 | NP_689571.2 | 6, 7 | 706 | ||
XM_011526436.2 | XP_011524738.1 | 6, 7 | 617 | |||
XM_011526437.2 | XP_011524739.1 | 5, 6 | 611 | |||
GenBank mRNA | AK131024.1 | - | 4, 5 | 600 | ||
AY358196.1 | - | 6, 7 | 620 | |||
BC029535.1 | - | 6, 7 | 587 |