Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF573 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172689.1 | NP_001166160.1 | 6 | 591 | ||
NM_001172690.1 | NP_001166161.1 | 5 | 400 | |||
NM_001172691.1 | NP_001166162.1 | 5 | 394 | |||
NM_001172692.1 | NP_001166163.1 | 4 | 267 | |||
NM_152360.3 | NP_689573.3 | 8 | 659 | |||
XM_011526446.2 | XP_011524748.1 | 6 | 478 | |||
XM_017026275.1 | XP_016881764.1 | 6 | 585 | |||
XM_017026276.1 | XP_016881765.1 | 6 | 1307 | |||
XM_017026277.1 | XP_016881766.1 | 6 | 472 | |||
XM_017026278.1 | XP_016881767.1 | 4 | 345 | |||
XM_017026279.1 | XP_016881768.1 | 4 | 290 | |||
XM_017026280.1 | XP_016881769.1 | 5 | 536 | |||
XM_017026281.1 | XP_016881770.1 | 4 | 1129 | |||
GenBank mRNA | AK055483.1 | - | 3 | 409 | ||
AK074539.1 | - | 6 | 587 | |||
BC029468.2 | - | 348 | ||||
BC042170.2 | - | 4 | 354 | 5014852 | ||
BC064962.1 | - | 8 | 629 | 6141400 | ||
BG572864.1 | - | 5 | 378 | |||
BP274132.1 | - | 5 | 396 | |||
DA778380.1 | - | 5 | 386 | |||
DB121870.1 | - | 4 | 240 |