Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF792 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175872.4 | NP_787068.3 | 4 | 2225 | ||
GenBank mRNA | AK095770.1 | - | 1 | 617 | ||
AK130901.1 | - | 1 | 934 | |||
AK304550.1 | - | 4 | 2140 | |||
BC101118.1 | - | 2 | 1621 | 40027074 | ||
BC101119.2 | - | 2 | 1621 | 40027075 | ||
BC101120.1 | - | 2 | 1621 | 40027081 |