Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCSAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145257.4 | NP_660300.3 | 3, 4 | 718 | ||
GenBank mRNA | AK123465.1 | - | 2, 3 | 507 | ||
AK291437.1 | - | 2, 3 | 679 | |||
BC015419.1 | - | 2, 3 | 692 | 4396825 | ||
BC060777.1 | - | 3, 4 | 1623 |