Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007255.2 | NP_001007256.1 | 3 | 1022 | ||
NM_152366.4 | NP_689579.3 | 3 | 1003 | |||
NR_033385.1 | 4 | 772 | ||||
NR_033386.1 | 4 | 791 | ||||
GenBank mRNA | AK093268.1 | - | 1 | 1367 | ||
AY601914.1 | - | 3 | 1021 | |||
BC022077.1 | - | 3 | 1002 | 4822843 | ||
BC032482.1 | - | 1 | 1396 | 5222141 | ||
BC066896.1 | - | 3 | 997 | 4828667 | ||
BU173744.1 | - | 4 | 768 | |||
BU622692.1 | - | 2 | 330 |