Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC44A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114106.2 | NP_001107578.1 | 13 | 1616 | ||
NM_001258340.1 | NP_001245269.1 | 13 | 1613 | |||
NM_001258341.1 | NP_001245270.1 | 13 | 1517 | |||
NM_001258342.1 | NP_001245271.1 | 12 | 1508 | |||
NM_001258343.1 | NP_001245272.1 | 12 | 1412 | |||
NM_001301079.1 | NP_001288008.1 | 12 | 1455 | |||
NM_152369.4 | NP_689582.2 | 12 | 1551 | |||
XM_005270440.2 | XP_005270497.1 | 13 | 1511 | |||
XM_005270441.2 | XP_005270498.1 | 13 | 1553 | |||
XM_005270442.1 | XP_005270499.1 | 13 | 1607 | |||
XM_011540632.2 | XP_011538934.1 | 12 | 1480 | |||
XM_011540633.2 | XP_011538935.1 | 12 | 1318 | |||
XM_011540636.2 | XP_011538938.1 | 14 | 1546 | |||
XM_017000262.1 | XP_016855751.1 | 12 | 1490 | |||
XM_017000263.1 | XP_016855752.1 | 13 | 1550 | |||
XM_017000264.1 | XP_016855753.1 | 13 | 1604 | |||
XM_017000265.1 | XP_016855754.1 | 13 | 1511 | |||
GenBank mRNA | AK300193.1 | - | 12 | 1349 | ||
AK301288.1 | - | 13 | 1357 | |||
AK302381.1 | - | 12 | 1392 | |||
AK303906.1 | - | 12 | 1487 | |||
AK313574.1 | - | 13 | 1501 | |||
AK316123.1 | - | 15 | 1458 | |||
AY358659.1 | - | 9 | 1070 | |||
BC033858.1 | - | 13 | 1579 | |||
BC053877.1 | - | 12 | 1474 | 5760394 | ||
BX648225.1 | - | 12 | 1480 |