Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152374.1 | NP_689587.1 | 2 | 675 | ||
GenBank mRNA | AK096303.1 | - | 2 | 675 | ||
AK096466.1 | - | 1 | 137 | |||
AK294352.1 | - | 2 | 308 | |||
BC026909.2 | - | 2 | 294 | 4932944 |