Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA5B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039500.2 | NP_001034589.2 | 22 | 3654 | ||
XM_006710360.3 | XP_006710423.2 | 19 | 2929 | |||
XM_011540683.2 | XP_011538985.1 | 22 | 3616 | |||
XM_011540684.2 | XP_011538986.1 | 22 | 3741 | |||
XM_011540685.2 | XP_011538987.1 | 22 | 3753 | |||
XM_011540688.2 | XP_011538990.1 | 21 | 3541 | |||
XM_011540690.2 | XP_011538992.1 | 21 | 3439 | |||
XM_011540691.2 | XP_011538993.1 | 21 | 3430 | |||
XM_011540692.2 | XP_011538994.1 | 22 | 3610 | |||
XM_011540697.2 | XP_011538999.1 | 16 | 2987 | |||
XM_017000305.1 | XP_016855794.1 | 16 | 2733 | |||
XM_017000306.1 | XP_016855795.1 | 17 | 2894 | |||
GenBank mRNA | AK057346.1 | - | 12 | 1938 | ||
AK295797.1 | - | 22 | 3638 | |||
BC101378.1 | - | 11 | 1734 | |||
BC101379.2 | - | 10 | 1659 | |||
BC101380.1 | - | 11 | 1734 | |||
BC101381.1 | - | 11 | 1734 |