Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000089.3 | NP_000080.2 | 51 | 4399 | ||
GenBank mRNA | AK226074.1 | - | 51 | 4050 | ||
AK297786.1 | - | 13 | 996 | |||
AK300194.1 | - | 10 | 621 | |||
AL833478.1 | - | 8 | 4706 | |||
BC042586.1 | - | 51 | 4046 | 4803351 | ||
BC054498.1 | - | 51 | 4028 | 6474549 | ||
J03464.1 | - | 51 | 4397 | |||
KU177988.1 | - | 51 | 3928 | |||
V00503.1 | - | 20 | 2051 | |||
X55525.1 | - | 8 | 1053 | |||
Z74616.1 | - | 51 | 4067 |