Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR1-1HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302812.1 | NP_001289741.1 | 5 | 675 | ||
NM_178463.3 | NP_848558.1 | 4 | 1056 | |||
GenBank mRNA | AK056985.1 | - | 4 | 1072 | ||
BC110595.2 | - | 4 | 731 | 40022808 | ||
F35210.1 | - | 1 | 31 |