Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000093.4 | NP_000084.3 | 6 | 1275 | ||
NM_001278074.1 | NP_001265003.1 | 6 | 1275 | |||
XM_017014266.1 | XP_016869755.1 | 6 | 1290 | |||
GenBank mRNA | AB209864.1 | - | 5 | 754 | ||
AB371583.1 | - | 6 | 1275 | |||
D90279.1 | - | 6 | 1018 |