Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYW5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039693.2 | NP_001034782.1 | 3 | 525 | ||
NR_004862.1 | 3 | 506 | ||||
NR_109905.1 | 3 | 525 | ||||
NR_109906.1 | 3 | 506 | ||||
NR_109907.1 | 2 | 370 | ||||
GenBank mRNA | AK095272.1 | - | 2 | 134 | ||
AK293064.1 | - | 3 | 273 | |||
AK298977.1 | - | 3 | 269 | |||
BC025758.1 | - | 3 | 253 | |||
BC132835.1 | - | 3 | 289 | 40146857 | ||
BC136837.1 | - | 3 | 289 | 9020828 | ||
BX482572.1 | - | 3 | 279 | |||
BX647677.1 | - | 3 | 507 | |||
BX648265.1 | - | 2 | 4775 | |||
CN261486.1 | - | 3 | 238 | |||
HY182974.1 | - | 3 | 300 |