Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174898.2 | NP_777558.1 | 5 | 448 | ||
XM_005263869.2 | XP_005263926.1 | 6 | 548 | |||
XM_005263870.3 | XP_005263927.1 | 6 | 453 | |||
XM_005263871.4 | XP_005263928.1 | 6 | 584 | |||
XM_011510580.2 | XP_011508882.1 | 6 | 547 | |||
XM_017003311.1 | XP_016858800.1 | 4 | 714 | |||
XM_017003312.1 | XP_016858801.1 | 3 | 765 | |||
XM_017003313.1 | XP_016858802.1 | 4 | 432 | |||
GenBank mRNA | AY358988.1 | - | 6 | 515 | ||
BC029126.1 | - | 5 | 448 | 5184582 | ||
BG194505.1 | - | 5 | 411 |