Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138572.2 | NP_612639.2 | 3, 4 | 324 | ||
XM_005248849.2 | XP_005248906.1 | 3, 4 | 313 | |||
XM_011514296.1 | XP_011512598.1 | 3, 4 | 315 | |||
XM_017010241.1 | XP_016865730.1 | 3, 4 | 315 | |||
XM_017010242.1 | XP_016865731.1 | 3, 4 | 478 | |||
XM_017010243.1 | XP_016865732.1 | 3, 4 | 478 | |||
XM_017010244.1 | XP_016865733.1 | 3, 4 | 313 | |||
XM_017010245.1 | XP_016865734.1 | 3, 4 | 313 | |||
GenBank mRNA | AF465841.1 | - | 3, 4 | 298 | ||
AK057383.1 | - | 3, 4 | 315 | |||
AK303097.1 | - | 3, 4 | 482 | |||
AL833401.1 | - | 3, 4 | 314 | |||
BC119678.1 | - | 3, 4 | 315 | 40116287 | ||
BC119679.1 | - | 3, 4 | 315 | 40116290 | ||
BI828714.1 | - | 3, 4 | 322 |