Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL16A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001856.3 | NP_001847.3 | 58 | 4010 | ||
XM_005270481.1 | XP_005270538.1 | 57 | 3957 | |||
XM_011540722.1 | XP_011539024.1 | 57 | 3960 | |||
XM_011540723.1 | XP_011539025.1 | 56 | 3912 | |||
XM_011540724.1 | XP_011539026.1 | 57 | 3882 | |||
XM_011540726.2 | XP_011539028.1 | 53 | 3428 | |||
XM_011540727.2 | XP_011539029.1 | 48 | 2766 | |||
XM_011540728.2 | XP_011539030.1 | 36 | 2248 | |||
XM_017000338.1 | XP_016855827.1 | 57 | 3972 | |||
XM_017000339.1 | XP_016855828.1 | 57 | 3969 | |||
XM_017000340.1 | XP_016855829.1 | 35 | 2211 | |||
XM_017000341.1 | XP_016855830.1 | 35 | 2001 | |||
GenBank mRNA | AB209571.1 | - | 52 | 3224 | ||
AF370368.1 | - | 2 | 892 | |||
M92642.1 | - | 59 | 3795 | |||
S57132.1 | - | 41 | 2391 |