Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYZL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304386.1 | NP_001291315.1 | 3, 4 | 553 | ||
NM_144634.3 | NP_653235.1 | 3, 4 | 572 | |||
XM_011533355.2 | XP_011531657.1 | 3, 4 | 615 | |||
XM_017005705.1 | XP_016861194.1 | 5, 6 | 927 | |||
XM_017005706.1 | XP_016861195.1 | 4, 5 | 685 | |||
GenBank mRNA | AF326749.1 | - | 3, 4 | 501 | ||
BC016747.2 | - | 3, 4 | 543 | 4830396 |