Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OCIAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014446.2 | NP_001014446.1 | 6 | 544 | ||
NM_001286774.1 | NP_001273703.1 | 5 | 447 | |||
GenBank mRNA | AK298305.1 | - | 6 | 513 | ||
AY472117.1 | - | 6 | 546 | |||
BG748993.1 | - | 6 | 449 | |||
BI546814.1 | - | 6 | 549 | |||
BM782144.1 | - | 6 | 514 | |||
BU508058.1 | - | 5 | 335 | |||
CR749310.1 | - | 5 | 1605 |