Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX4I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318786.1 | NP_001305715.1 | 3 | 325 | ||
NM_001318788.1 | NP_001305717.1 | 3 | 304 | |||
NM_001318794.1 | NP_001305723.1 | 3 | 304 | |||
NM_001318797.1 | NP_001305726.1 | 3 | 169 | |||
NM_001318802.1 | NP_001305731.1 | 3 | 304 | |||
NM_001861.4 | NP_001852.1 | 3 | 304 | |||
GenBank mRNA | AK027136.1 | - | 1 | 1926 | ||
AK311847.1 | - | 3 | 176 | |||
BC008704.1 | - | 3 | 162 | 2990066 | ||
BC021236.2 | - | 3 | 155 | 4309698 | ||
BC047869.1 | - | 3 | 249 | |||
BC062437.1 | - | 3 | 183 | 6451700 | ||
BE615615.1 | - | 2 | 121 | |||
BE728651.1 | - | 3 | 166 | |||
BG614492.1 | - | 3 | 189 | |||
BM835194.1 | - | 3 | 280 | |||
BP343428.1 | - | 3 | 182 | |||
BT019825.1 | - | 2 | 117 | |||
BU599266.1 | - | 163 | ||||
CD677397.1 | - | 3 | 163 | |||
CN406213.1 | - | 3 | 169 | |||
DA623889.1 | - | 3 | 306 | |||
DB502762.1 | - | 3 | 169 | |||
M21575.1 | - | 3 | 151 | |||
M34600.1 | - | 2 | 117 | |||
U90915.1 | - | 3 | 187 | |||
X54802.1 | - | 3 | 179 |