Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206919.1 | NP_996802.1 | 4 | 683 | ||
XM_005265719.3 | XP_005265776.1 | 4 | 797 | |||
XM_005265720.3 | XP_005265777.1 | 4 | 865 | |||
XM_006714036.3 | XP_006714099.1 | 4 | 557 | |||
GenBank mRNA | AY439003.1 | - | 4 | 683 |