Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198449.2 | NP_940851.1 | 4, 5 | 717 | ||
XM_011543146.2 | XP_011541448.1 | 5, 6 | 499 | |||
GenBank mRNA | AK300860.1 | - | 4, 5 | 682 | ||
AK304226.1 | - | 4, 5 | 588 | |||
BC059398.1 | - | 4, 5 | 684 | 30332712 |
Gene Symbol: EMBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003955.1 | 3, 4 | 620 | |||
GenBank mRNA | AK128714.1 | - | 3, 4 | 620 |