Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MROH2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173489.4 | NP_775760.3 | 6, 7 | 1107 | ||
XM_011513952.1 | XP_011512254.1 | 6, 7 | 1093 | |||
XM_011513953.1 | XP_011512255.1 | 5, 6 | 1031 | |||
GenBank mRNA | AL832763.1 | - | 1, 2 | 32 | ||
BC034600.1 | - | 6, 7 | 1071 | |||
BC052962.1 | - | 6, 7 | 1068 | 4838071 | ||
CR749346.1 | - | 1, 2 | 22 |