Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102609.2 | NP_001096079.1 | 2 | 159 | ||
NM_001305393.1 | NP_001292322.1 | 2 | 178 | |||
NM_001305394.1 | NP_001292323.1 | 2 | 167 | |||
NM_001305395.1 | NP_001292324.1 | 2 | 159 | |||
NM_001305396.1 | NP_001292325.1 | 2 | 148 | |||
NM_001305397.1 | NP_001292326.1 | 2 | 159 | |||
NR_131091.1 | 2 | 159 | ||||
XM_011534432.2 | XP_011532734.1 | 2 | 630 | |||
XM_017009029.1 | XP_016864518.1 | 2 | 1346 | |||
XM_017009030.1 | XP_016864519.1 | 2 | 1345 | |||
XM_017009031.1 | XP_016864520.1 | 2 | 219 | |||
XM_017009032.1 | XP_016864521.1 | 2 | 143 | |||
XM_017009033.1 | XP_016864522.1 | 2 | 210 | |||
GenBank mRNA | AA758527.1 | - | 3 | 381 | ||
BC030767.1 | - | 2 | 139 | |||
BC092511.1 | - | 1 | 1139 | |||
BU561730.1 | - | 2 | 95 | |||
BX100176.1 | - | 1 | 79 | |||
HY000873.1 | - | 1 | 71 | |||
HY001392.1 | - | 1 | 82 | |||
HY005987.1 | - | 2 | 178 | |||
HY016595.1 | - | 2 | 84 | |||
HY038777.1 | - | 2 | 159 |