Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX6B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001863.4 | NP_001854.1 | 4 | 425 | ||
GenBank mRNA | AK312140.1 | - | 4 | 322 | ||
BC001015.2 | - | 4 | 395 | 3344701 | ||
BC002478.2 | - | 4 | 294 | 3347455 | ||
BT006945.1 | - | 3 | 233 | |||
CR456789.1 | - | 3 | 233 | |||
CR542137.1 | - | 3 | 233 | |||
X13923.1 | - | 4 | 277 | |||
X54473.1 | - | 4 | 280 |