Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX7A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001865.3 | NP_001856.2 | 1, 2 | 207 | ||
NR_029466.1 | 1, 2 | 207 | ||||
GenBank mRNA | AK312154.1 | - | 1, 2 | 62 | ||
BC101826.1 | - | 1, 2 | 50 | 8069332 | ||
BC101828.1 | - | 1, 2 | 50 | 8069334 | ||
CB139775.1 | - | 1, 2 | 66 | |||
CK821341.1 | - | 1, 2 | 218 | |||
CR407646.1 | - | 1, 2 | 13 | |||
CR542125.1 | - | 1, 2 | 13 | |||
HE647861.1 | - | 1, 2 | 207 | |||
X15822.1 | - | 1, 2 | 26 |